Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAAGGCTGGGGGATGCCAAAGGT[G/T]CGGAAGGCAGGGTGAGGCTCGGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603500 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
B3GNT9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
B3GNT9 - UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033309.2 | 1332 | Silent Mutation | NP_171608.2 |
C16orf70 - chromosome 16 open reading frame 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320540.1 | 1332 | Intron | NP_001307469.1 | |||
NM_001320541.1 | 1332 | Intron | NP_001307470.1 | |||
NM_001320542.1 | 1332 | Intron | NP_001307471.1 | |||
NM_001320543.1 | 1332 | Intron | NP_001307472.1 | |||
NM_025187.4 | 1332 | Intron | NP_079463.2 | |||
XM_006721284.3 | 1332 | Intron | XP_006721347.1 | |||
XM_011523350.2 | 1332 | Intron | XP_011521652.1 | |||
XM_017023729.1 | 1332 | Intron | XP_016879218.1 | |||
XM_017023730.1 | 1332 | Intron | XP_016879219.1 | |||
XM_017023731.1 | 1332 | Intron | XP_016879220.1 |
TRADD - TNFRSF1A associated via death domain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |