Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGCAGCGTTTCCGGTACAGGTC[A/G]AACTTGCCAAAGCTGATGTAGGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606007 MIM: 614403 | ||||||||||||||||||||
Literature Links: |
POLR3K PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POLR3K - RNA polymerase III subunit K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHBDF1 - rhomboid 5 homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022450.3 | 2595 | Silent Mutation | TTC,TTT | F,F 795 | NP_071895.3 | |
XM_005255494.1 | 2595 | Silent Mutation | TTC,TTT | F,F 795 | XP_005255551.1 | |
XM_005255498.2 | 2595 | Silent Mutation | TTC,TTT | F,F 479 | XP_005255555.1 | |
XM_006720921.1 | 2595 | Silent Mutation | TTC,TTT | F,F 818 | XP_006720984.1 | |
XM_017023556.1 | 2595 | Silent Mutation | TTC,TTT | F,F 818 | XP_016879045.1 | |
XM_017023557.1 | 2595 | Silent Mutation | TTC,TTT | F,F 764 | XP_016879046.1 | |
XM_017023558.1 | 2595 | Intron | XP_016879047.1 |
SNRNP25 - small nuclear ribonucleoprotein U11/U12 subunit 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024571.3 | 2595 | Intron | NP_078847.1 |