Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGCCGTTCAGGCCTCGTGGCAGC[A/C]CCAGATCAAGGGCGAGGTTGCGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605294 | ||||||||||||||||||||
Literature Links: |
CHST6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHST6 - carbohydrate sulfotransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021615.4 | 1919 | Missense Mutation | GGG,GTG | G,V 375 | NP_067628.1 | |
XM_005255955.4 | 1919 | Missense Mutation | GGG,GTG | G,V 375 | XP_005256012.1 | |
XM_011523085.2 | 1919 | Missense Mutation | GGG,GTG | G,V 375 | XP_011521387.1 |