Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATATGGCATCTGCCAGCTCTAGC[C/T]GGGCAGGAGTGGCCCTGCCTTTTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602308 | ||||||||||||||||||||
Literature Links: |
WWP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WWP2 - WW domain containing E3 ubiquitin protein ligase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270453.1 | 145 | Intron | NP_001257382.1 | |||
NM_001270454.1 | 145 | Missense Mutation | CGG,TGG | R,W 8 | NP_001257383.1 | |
NM_001270455.1 | 145 | Missense Mutation | CGG,TGG | R,W 8 | NP_001257384.1 | |
NM_007014.4 | 145 | Missense Mutation | CGG,TGG | R,W 8 | NP_008945.2 | |
NM_199424.2 | 145 | Intron | NP_955456.1 | |||
XM_011522823.2 | 145 | Missense Mutation | CGG,TGG | R,W 8 | XP_011521125.1 | |
XM_011522825.1 | 145 | Missense Mutation | CGG,TGG | R,W 8 | XP_011521127.1 | |
XM_011522826.2 | 145 | Intron | XP_011521128.1 | |||
XM_017022879.1 | 145 | Missense Mutation | CGG,TGG | R,W 8 | XP_016878368.1 | |
XM_017022880.1 | 145 | Missense Mutation | CGG,TGG | R,W 8 | XP_016878369.1 | |
XM_017022881.1 | 145 | Missense Mutation | CGG,TGG | R,W 8 | XP_016878370.1 |