Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAAGCAATCCTCCCTCAGCTTCC[C/T]GTTTCTGAGGTTACAGGTATGCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616320 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAHD1 - fumarylacetoacetate hydrolase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018104.2 | 745 | Missense Mutation | CGT,TGT | R,C 247 | NP_001018114.1 | |
NM_001142398.1 | 745 | Intron | NP_001135870.1 | |||
NM_031208.3 | 745 | Intron | NP_112485.1 |
MEIOB - meiosis specific with OB domains | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163560.2 | 745 | Intron | NP_001157032.1 | |||
NM_152764.2 | 745 | Intron | NP_689977.2 |