Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGGGTGTAGGGCCAGGGCGCCA[C/T]GAGGAGCGGAGGTCAGCGGGGATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610087 MIM: 605777 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRMT7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMPD3 - sphingomyelin phosphodiesterase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018667.3 | 4287 | UTR 3 | NP_061137.1 | |||
XM_005256031.3 | 4287 | UTR 3 | XP_005256088.1 | |||
XM_005256032.3 | 4287 | UTR 3 | XP_005256089.1 | |||
XM_011523207.1 | 4287 | UTR 3 | XP_011521509.1 | |||
XM_011523208.2 | 4287 | UTR 3 | XP_011521510.1 | |||
XM_011523209.2 | 4287 | UTR 3 | XP_011521511.1 | |||
XM_011523210.2 | 4287 | Intron | XP_011521512.1 | |||
XM_017023405.1 | 4287 | UTR 3 | XP_016878894.1 | |||
XM_017023406.1 | 4287 | UTR 3 | XP_016878895.1 | |||
XM_017023407.1 | 4287 | UTR 3 | XP_016878896.1 | |||
XM_017023408.1 | 4287 | UTR 3 | XP_016878897.1 |