Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAATGCAAATACTTGTGCACTGGG[A/T]GAGGTGAGCCGTTTTGCAGGCTGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614094 | ||||||||||||||||||||
Literature Links: |
MARVELD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MARVELD3 - MARVEL domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017967.3 | 506 | Nonsense Mutation | AGA,TGA | R,* 198 | NP_001017967.2 | |
NM_001271329.1 | 506 | Silent Mutation | GGA,GGT | G,G 143 | NP_001258258.1 | |
NM_052858.5 | 506 | Nonsense Mutation | AGA,TGA | R,* 198 | NP_443090.4 | |
XM_011523449.2 | 506 | Nonsense Mutation | AGA,TGA | R,* 198 | XP_011521751.1 |