Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAATCAGATGGCCATCAGCAACAA[C/T]AGCCAACAGAGCAATGAGCAGACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606151 MIM: 615857 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BBS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BBS2 - Bardet-Biedl syndrome 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031885.3 | 1395 | Intron | NP_114091.3 | |||
XM_005256080.2 | 1395 | Intron | XP_005256137.1 |
OGFOD1 - 2-oxoglutarate and iron dependent oxygenase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324357.1 | 1395 | Silent Mutation | AAC,AAT | N,N 408 | NP_001311286.1 | |
NM_001324358.1 | 1395 | Silent Mutation | AAC,AAT | N,N 354 | NP_001311287.1 | |
NM_001324359.1 | 1395 | Silent Mutation | AAC,AAT | N,N 312 | NP_001311288.1 | |
NM_001324360.1 | 1395 | Silent Mutation | AAC,AAT | N,N 312 | NP_001311289.1 | |
NM_001324361.1 | 1395 | Silent Mutation | AAC,AAT | N,N 354 | NP_001311290.1 | |
NM_001324362.1 | 1395 | Silent Mutation | AAC,AAT | N,N 269 | NP_001311291.1 | |
NM_001324363.1 | 1395 | Silent Mutation | AAC,AAT | N,N 371 | NP_001311292.1 | |
NM_018233.3 | 1395 | Silent Mutation | AAC,AAT | N,N 409 | NP_060703.3 |