Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGCCCAGGTCTTGTCCTGGATCA[C/T]GCAAGTGAATTAGAACCTCAAATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TEKT5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TEKT5 - tektin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144674.1 | Intron | NP_653275.1 | ||||
XM_011522383.2 | Intron | XP_011520685.1 | ||||
XM_011522385.2 | Intron | XP_011520687.1 | ||||
XM_011522389.1 | Intron | XP_011520691.1 | ||||
XM_011522390.2 | Intron | XP_011520692.1 | ||||
XM_017022955.1 | Intron | XP_016878444.1 | ||||
XM_017022956.1 | Intron | XP_016878445.1 | ||||
XM_017022957.1 | Intron | XP_016878446.1 | ||||
XM_017022958.1 | Intron | XP_016878447.1 |