Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTAAGAAACGTGCCCGGCGGATC[C/T]TGCAGGCTTGTGGCGGGAACAGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138210 | ||||||||||||||||||||
Literature Links: |
GPT2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPT2 - glutamic--pyruvic transaminase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142466.2 | 501 | Silent Mutation | CTG,TTG | L,L 39 | NP_001135938.1 | |
NM_133443.3 | 501 | Silent Mutation | CTG,TTG | L,L 139 | NP_597700.1 | |
XM_017023790.1 | 501 | Intron | XP_016879279.1 |