Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACACAATGAGATCATGGCCATATT[C/G]TTTACCAGTGGAACAAGTGGATATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 145505 | ||||||||||||||||||||
Literature Links: |
ACSM3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACSM3 - acyl-CoA synthetase medium-chain family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005622.3 | 880 | Missense Mutation | TTC,TTG | F,L 233 | NP_005613.2 | |
NM_202000.2 | 880 | Missense Mutation | TTC,TTG | F,L 233 | NP_973729.1 | |
XM_011545911.1 | 880 | Missense Mutation | TTC,TTG | F,L 270 | XP_011544213.1 | |
XM_017023523.1 | 880 | Missense Mutation | TTC,TTG | F,L 233 | XP_016879012.1 | |
XM_017023524.1 | 880 | Missense Mutation | TTC,TTG | F,L 233 | XP_016879013.1 |
ERI2 - ERI1 exoribonuclease family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142725.1 | 880 | Intron | NP_001136197.1 | |||
NM_080663.2 | 880 | Intron | NP_542394.2 |