Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGCAGCTGGCATGGAACCGTCC[A/G]TCTCAGCGGCTGCTTGGTGGTGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C16orf89 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C16orf89 - chromosome 16 open reading frame 89 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098514.2 | 1247 | UTR 3 | NP_001091984.2 | |||
NM_152459.4 | 1247 | UTR 3 | NP_689672.4 | |||
XM_005255143.3 | 1247 | Intron | XP_005255200.2 | |||
XM_011522392.2 | 1247 | UTR 3 | XP_011520694.1 | |||
XM_017022972.1 | 1247 | UTR 3 | XP_016878461.1 | |||
XM_017022973.1 | 1247 | Intron | XP_016878462.1 | |||
XM_017022974.1 | 1247 | Intron | XP_016878463.1 |
NAGPA-AS1 - NAGPA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |