Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACCGGCCACTGGCCTCACTGAAA[C/T]GCTCTTCCCCCTGTCTCCTGAGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C16orf62 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C16orf62 - chromosome 16 open reading frame 62 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IQCK - IQ motif containing K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KNOP1 - lysine rich nucleolar protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012991.2 | Intron | NP_001013009.2 | ||||
XM_005255317.4 | Intron | XP_005255374.1 | ||||
XM_006721046.3 | Intron | XP_006721109.1 | ||||
XM_011545855.1 | Intron | XP_011544157.1 | ||||
XM_017023231.1 | Intron | XP_016878720.1 | ||||
XM_017023232.1 | Intron | XP_016878721.1 |