Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTTCCCCCATGAGCCCTGGCTGC[A/G]GCTGTGCCGGGAGAGAGGCGGCACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605754 | ||||||||||||||||||||
Literature Links: |
LOC107987233 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC107987233 - uncharacterized LOC107987233 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NHLRC4 - NHL repeat containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGQ - phosphatidylinositol glycan anchor biosynthesis class Q | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004204.3 | 422 | Missense Mutation | CAG,CGG | Q,R 95 | NP_004195.2 | |
NM_148920.2 | 422 | Missense Mutation | CAG,CGG | Q,R 95 | NP_683721.1 |
PRR35 - proline rich 35 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |