Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCAAATTAGTGTCTATTTTGACA[A/G]TAACACCGCAGCATTAACAGAACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616787 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C16orf90 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C16orf90 - chromosome 16 open reading frame 90 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CLUAP1 - clusterin associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015041.2 | Intron | NP_055856.1 | ||||
NM_024793.2 | Intron | NP_079069.1 | ||||
XM_006720867.3 | Intron | XP_006720930.1 | ||||
XM_017023068.1 | Intron | XP_016878557.1 | ||||
XM_017023069.1 | Intron | XP_016878558.1 | ||||
XM_017023070.1 | Intron | XP_016878559.1 | ||||
XM_017023071.1 | Intron | XP_016878560.1 | ||||
XM_017023072.1 | Intron | XP_016878561.1 | ||||
XM_017023073.1 | Intron | XP_016878562.1 |