Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGAGATTCACCTGAGACCAAAT[G/T]GTTCCAATAAATATCTATGATCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601364 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDH13 - cadherin 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001220488.1 | Intron | NP_001207417.1 | ||||
NM_001220489.1 | Intron | NP_001207418.1 | ||||
NM_001220490.1 | Intron | NP_001207419.1 | ||||
NM_001220491.1 | Intron | NP_001207420.1 | ||||
NM_001220492.1 | Intron | NP_001207421.1 | ||||
NM_001257.4 | Intron | NP_001248.1 | ||||
XM_011522804.2 | Intron | XP_011521106.1 | ||||
XM_017022848.1 | Intron | XP_016878337.1 | ||||
XM_017022849.1 | Intron | XP_016878338.1 |