Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACACAAGCCCTGCAGGGCTTCCT[C/G]TTTCTCCTCTTCCTCCCGCTGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609899 MIM: 614578 MIM: 602474 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KREMEN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KREMEN2 - kringle containing transmembrane protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253725.1 | 335 | Silent Mutation | CTC,CTG | L,L 10 | NP_001240654.1 | |
NM_001253726.1 | 335 | Silent Mutation | CTC,CTG | L,L 10 | NP_001240655.1 | |
NM_024507.3 | 335 | Silent Mutation | CTC,CTG | L,L 10 | NP_078783.1 | |
NM_172229.2 | 335 | Silent Mutation | CTC,CTG | L,L 10 | NP_757384.1 |
PAQR4 - progestin and adipoQ receptor family member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PKMYT1 - protein kinase, membrane associated tyrosine/threonine 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |