Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGCCATCAAATCCACCACGCTGC[A/G]GGTTGACCAGTCCATCCTGACAGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 108730 | ||||||||||||||||||||
Literature Links: |
ATP2A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP2A1 - ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286075.1 | 705 | Missense Mutation | CAG,CGG | Q,R 49 | NP_001273004.1 | |
NM_004320.4 | 705 | Missense Mutation | CAG,CGG | Q,R 174 | NP_004311.1 | |
NM_173201.3 | 705 | Missense Mutation | CAG,CGG | Q,R 174 | NP_775293.1 |
ATP2A1-AS1 - ATP2A1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |