Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTACCTTTTCCACTTGTCTGAA[A/G]ACCCGCAGCAGGTTTCCACGAAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609926 MIM: 604119 | ||||||||||||||||||||
Literature Links: |
DPEP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPEP3 - dipeptidase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129758.1 | 1378 | Silent Mutation | GTC,GTT | V,V 428 | NP_001123230.1 | |
NM_022357.3 | 1378 | Silent Mutation | GTC,GTT | V,V 429 | NP_071752.3 |
SLC12A4 - solute carrier family 12 member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |