Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATATCTCCAGTGGCGATTTCCCA[A/G]AGGACGATTCCAAAGCTAAAAGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615153 MIM: 614151 | ||||||||||||||||||||
Literature Links: |
MLKL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MLKL - mixed lineage kinase domain-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142497.2 | 1904 | Missense Mutation | TCT,TTT | S,F 184 | NP_001135969.1 | |
NM_152649.3 | 1904 | Silent Mutation | CTC,CTT | L,L 402 | NP_689862.1 | |
XM_005255834.3 | 1904 | Silent Mutation | CTC,CTT | L,L 402 | XP_005255891.1 | |
XM_011522936.2 | 1904 | Intron | XP_011521238.1 |
RFWD3 - ring finger and WD repeat domain 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |