Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAAATGAACCAGAAGACAATCCT[C/T]GTGCTCCTCATTCTGGCCGTCATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602268 MIM: 603735 MIM: 605129 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AOC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AOC2 - amine oxidase, copper containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001158.3 | 242 | Intron | NP_001149.2 | |||
NM_009590.2 | 242 | Intron | NP_033720.2 |
AOC3 - amine oxidase, copper containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277731.1 | 242 | Silent Mutation | CTC,CTT | L,L 7 | NP_001264660.1 | |
NM_001277732.1 | 242 | Intron | NP_001264661.1 | |||
NM_003734.3 | 242 | Silent Mutation | CTC,CTT | L,L 7 | NP_003725.1 | |
XM_011525419.1 | 242 | Silent Mutation | CTC,CTT | L,L 7 | XP_011523721.1 | |
XM_011525420.2 | 242 | Silent Mutation | CTC,CTT | L,L 7 | XP_011523722.1 |
PSME3 - proteasome activator subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |