Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCTTGGAAAAGGGGCCCGGGGC[C/T]AAGGGACTGTCGCTTTCCAGGTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614966 MIM: 608613 | ||||||||||||||||||||
Literature Links: |
SCRN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SCRN2 - secernin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SP6 - Sp6 transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258248.1 | 502 | Silent Mutation | TTA,TTG | L,L 87 | NP_001245177.1 | |
NM_199262.2 | 502 | Silent Mutation | TTA,TTG | L,L 87 | NP_954871.1 | |
XM_006722115.3 | 502 | Silent Mutation | TTA,TTG | L,L 87 | XP_006722178.1 |