Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCACGGCGAAAAAGAACTCCGG[A/G]AAGAAGGTGCGGACGTACACGGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616161 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHRS7C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHRS7C - dehydrogenase/reductase 7C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105571.2 | 880 | Silent Mutation | TTC,TTT | F,F 289 | NP_001099041.1 | |
NM_001220493.1 | 880 | Silent Mutation | TTC,TTT | F,F 290 | NP_001207422.1 |