Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGCATAAAGCATGCTTCCATTG[C/T]GAGACCTGCAAGATGACACTGAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602920 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LASP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LASP1 - LIM and SH3 protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271608.1 | 427 | Nonsense Mutation | CGA,TGA | R,* 5 | NP_001258537.1 | |
NM_006148.3 | 427 | Silent Mutation | TGC,TGT | C,C 32 | NP_006139.1 |