Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCACCACTTTCACGTCTTTCAAC[G/T]CTTTTTATCTTTTTGTTTTATAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109270 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC4A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC4A1 - solute carrier family 4 member 1 (Diego blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000342.3 | 4112 | UTR 3 | NP_000333.1 | |||
XM_005257593.4 | 4112 | UTR 3 | XP_005257650.1 | |||
XM_011525129.1 | 4112 | UTR 3 | XP_011523431.1 | |||
XM_011525130.1 | 4112 | Intron | XP_011523432.1 |