Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGCTCTTTGCTGCTGAACGTGTT[A/C]AGGTAGTGCACGAAAGGCGGCTCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615124 MIM: 606779 | ||||||||||||||||||||
Literature Links: |
ANKRD13B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD13B - ankyrin repeat domain 13B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CORO6 - coronin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032854.3 | 860 | Silent Mutation | CTG,CTT | L,L 306 | NP_116243.2 | |
XM_005258047.3 | 860 | Silent Mutation | CTG,CTT | L,L 306 | XP_005258104.1 | |
XM_005258048.4 | 860 | Silent Mutation | CTG,CTT | L,L 377 | XP_005258105.2 | |
XM_005258056.4 | 860 | Silent Mutation | CTG,CTT | L,L 72 | XP_005258113.1 | |
XM_011525385.2 | 860 | Silent Mutation | CTG,CTT | L,L 377 | XP_011523687.1 | |
XM_011525386.2 | 860 | Silent Mutation | CTG,CTT | L,L 342 | XP_011523688.1 | |
XM_011525387.2 | 860 | Silent Mutation | CTG,CTT | L,L 336 | XP_011523689.1 | |
XM_011525388.2 | 860 | Silent Mutation | CTG,CTT | L,L 377 | XP_011523690.1 | |
XM_011525389.2 | 860 | Intron | XP_011523691.1 | |||
XM_011525390.2 | 860 | Silent Mutation | CTG,CTT | L,L 72 | XP_011523692.1 | |
XM_011525391.2 | 860 | Silent Mutation | CTG,CTT | L,L 72 | XP_011523693.1 | |
XM_017025235.1 | 860 | Silent Mutation | CTG,CTT | L,L 265 | XP_016880724.1 |
SSH2 - slingshot protein phosphatase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |