Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACTACTTGGTGAAATGACTTCT[C/T]GTCACATTGCTGAAGAACATATACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608034 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASPA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASPA - aspartoacylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000049.2 | 317 | Missense Mutation | CGT,TGT | R,C 4 | NP_000040.1 | |
NM_001128085.1 | 317 | Missense Mutation | CGT,TGT | R,C 4 | NP_001121557.1 | |
XM_017024661.1 | 317 | Missense Mutation | CGT,TGT | R,C 4 | XP_016880150.1 |
SPATA22 - spermatogenesis associated 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170695.1 | 317 | Intron | NP_001164166.1 | |||
NM_001170696.1 | 317 | Intron | NP_001164167.1 | |||
NM_001170697.1 | 317 | Intron | NP_001164168.1 | |||
NM_001170698.1 | 317 | Intron | NP_001164169.1 | |||
NM_001170699.1 | 317 | Intron | NP_001164170.1 | |||
NM_001321336.1 | 317 | Intron | NP_001308265.1 | |||
NM_001321337.1 | 317 | Intron | NP_001308266.1 | |||
NM_032598.4 | 317 | Intron | NP_115987.2 |