Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCCCATCCTCAGGGTCCTGCCCA[C/T]TGGGGGAGATGGGCTGACCTCCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600147 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEOX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MEOX1 - mesenchyme homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040002.1 | 1144 | Missense Mutation | AAT,AGT | N,S 123 | NP_001035091.1 | |
NM_004527.3 | 1144 | Missense Mutation | AAT,AGT | N,S 238 | NP_004518.1 | |
NM_013999.3 | 1144 | Silent Mutation | CAA,CAG | Q,Q 180 | NP_054705.1 | |
XM_011524818.1 | 1144 | UTR 3 | XP_011523120.1 |