Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATGGTGACCATGGTCACAGGGTT[C/G]AGCACGGTCTGGCCCACCAGCTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603039 MIM: 611418 | ||||||||||||||||||||
Literature Links: |
MNT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MNT - MAX network transcriptional repressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020310.2 | 975 | Silent Mutation | CTC,CTG | L,L 563 | NP_064706.1 | |
XM_011523868.2 | 975 | Silent Mutation | CTC,CTG | L,L 571 | XP_011522170.1 | |
XM_011523869.1 | 975 | Silent Mutation | CTC,CTG | L,L 282 | XP_011522171.1 | |
XM_017024654.1 | 975 | Silent Mutation | CTC,CTG | L,L 282 | XP_016880143.1 |
SGSM2 - small G protein signaling modulator 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |