Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCACAGCCTGTTCAACCTCAGCA[A/G]GGTTATATTCCTCCAATGGCACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603428 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF207 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF207 - zinc finger protein 207 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032293.2 | 688 | Intron | NP_001027464.1 | |||
NM_001098507.1 | 688 | Intron | NP_001091977.1 | |||
NM_003457.3 | 688 | Intron | NP_003448.1 | |||
XM_011525185.2 | 688 | Silent Mutation | CAA,CAG | Q,Q 82 | XP_011523487.1 | |
XM_017025019.1 | 688 | Silent Mutation | CAA,CAG | Q,Q 136 | XP_016880508.1 | |
XM_017025020.1 | 688 | Intron | XP_016880509.1 |