Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGATGCTGCCTTCCACATCACGCC[A/G]CCTCAACCACTATGTTCTGTATAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611218 | ||||||||||||||||||||
Literature Links: |
GSDMA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GSDMA - gasdermin A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMD3 - proteasome 26S subunit, non-ATPase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002809.3 | 523 | Missense Mutation | CAC,CGC | H,R 107 | NP_002800.2 |