Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTCGGCCAAGACCCCATCCTGC[A/G]GCAGGCTGGCATCGCCTTTGGGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616157 MIM: 131560 | ||||||||||||||||||||
Literature Links: |
DHRS13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHRS13 - dehydrogenase/reductase 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FLOT2 - flotillin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHF12 - PHD finger protein 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033561.1 | 2775 | Missense Mutation | CCG,CTG | P,L 979 | NP_001028733.1 | |
NM_001290131.1 | 2775 | Intron | NP_001277060.1 | |||
NM_020889.2 | 2775 | Intron | NP_065940.1 | |||
XM_005258015.3 | 2775 | Missense Mutation | CCG,CTG | P,L 889 | XP_005258072.1 | |
XM_005258016.3 | 2775 | Missense Mutation | CCG,CTG | P,L 762 | XP_005258073.1 | |
XM_011525078.2 | 2775 | Missense Mutation | CCG,CTG | P,L 957 | XP_011523380.1 | |
XM_011525079.2 | 2775 | Missense Mutation | CCG,CTG | P,L 946 | XP_011523381.1 | |
XM_011525082.2 | 2775 | Intron | XP_011523384.1 | |||
XM_011525083.2 | 2775 | Intron | XP_011523385.1 | |||
XM_017024903.1 | 2775 | Missense Mutation | CCG,CTG | P,L 997 | XP_016880392.1 | |
XM_017024904.1 | 2775 | Missense Mutation | CCG,CTG | P,L 948 | XP_016880393.1 |