Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAGGAGGACAACAGCTACAACCG[C/G]GGGTAACGAAATCCTCGGAGTCCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 160993 | ||||||||||||||||||||
Literature Links: |
DCAKD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCAKD - dephospho-CoA kinase domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128631.2 | 160 | Intron | NP_001122103.1 | |||
NM_001288654.2 | 160 | Intron | NP_001275583.1 | |||
NM_001288655.1 | 160 | Intron | NP_001275584.1 | |||
NM_001321326.1 | 160 | Intron | NP_001308255.1 | |||
NM_024819.6 | 160 | Intron | NP_079095.3 | |||
XM_005257688.2 | 160 | Intron | XP_005257745.1 | |||
XM_011525262.2 | 160 | Intron | XP_011523564.1 | |||
XM_017025102.1 | 160 | Intron | XP_016880591.1 | |||
XM_017025103.1 | 160 | Intron | XP_016880592.1 | |||
XM_017025104.1 | 160 | Intron | XP_016880593.1 | |||
XM_017025105.1 | 160 | Intron | XP_016880594.1 | |||
XM_017025106.1 | 160 | Intron | XP_016880595.1 |
NMT1 - N-myristoyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021079.4 | 160 | Silent Mutation | CGC,CGG | R,R 43 | NP_066565.1 | |
XM_011524858.2 | 160 | Silent Mutation | CGC,CGG | R,R 43 | XP_011523160.1 |