Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTCAATCACTTTAATAGATGTC[C/T]ATAGGTAGTTCATATGAATGCTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608626 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC47 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC47 - coiled-coil domain containing 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020198.2 | 3388 | UTR 3 | NP_064583.2 | |||
XM_005257527.2 | 3388 | Intron | XP_005257584.1 |
STRADA - STE20-related kinase adaptor alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |