Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAACAGTGTAGATTACAGCAGGC[A/G]TTCTCTACCTGGACTCATGATTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606969 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DBIL5P PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DBIL5P - diazepam binding inhibitor-like 5, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GEMIN4 - gem nuclear organelle associated protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GLOD4 - glyoxalase domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016080.3 | 1198 | UTR 3 | NP_057164.3 | |||
XM_006721540.2 | 1198 | UTR 3 | XP_006721603.1 | |||
XM_017024711.1 | 1198 | UTR 3 | XP_016880200.1 | |||
XM_017024712.1 | 1198 | UTR 3 | XP_016880201.1 |