Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCTTCTGAACTTCATTAGTGAC[C/T]TTGTGAATGATAAGTTCAATAGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609801 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD300E PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD300E - CD300e molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181449.2 | 3292 | UTR 3 | NP_852114.2 | |||
XM_017024575.1 | 3292 | Intron | XP_016880064.1 |
LOC101928343 - uncharacterized LOC101928343 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |