Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGATGGAAGTGAGCACAGTCCT[C/T]GCCTCCACCCAGCCCGTGCCCCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605999 | ||||||||||||||||||||
Literature Links: |
CLEC10A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLEC10A - C-type lectin domain family 10 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005256412.1 | 1176 | Missense Mutation | AAG,GAG | K,E 253 | XP_005256469.1 | |
XM_011523613.1 | 1176 | Missense Mutation | AAG,GAG | K,E 283 | XP_011521915.1 | |
XM_011523614.2 | 1176 | Missense Mutation | AAG,GAG | K,E 283 | XP_011521916.1 | |
XM_011523615.1 | 1176 | Missense Mutation | AAG,GAG | K,E 279 | XP_011521917.1 |