Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTCATCTTCAGTGGTAGGGATA[C/T]TGCTGATGTCGATGTAAACCTGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607875 MIM: 606538 | ||||||||||||||||||||
Literature Links: |
INPP5K PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
INPP5K - inositol polyphosphate-5-phosphatase K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135642.1 | 1374 | Missense Mutation | AAT,AGT | N,S 326 | NP_001129114.1 | |
NM_016532.3 | 1374 | Missense Mutation | AAT,AGT | N,S 402 | NP_057616.2 | |
NM_130766.2 | 1374 | Missense Mutation | AAT,AGT | N,S 326 | NP_570122.1 | |
XM_005256683.2 | 1374 | Missense Mutation | AAT,AGT | N,S 326 | XP_005256740.1 | |
XM_005256685.1 | 1374 | Missense Mutation | AAT,AGT | N,S 310 | XP_005256742.1 | |
XM_005256686.1 | 1374 | Missense Mutation | AAT,AGT | N,S 310 | XP_005256743.1 | |
XM_011523934.1 | 1374 | Missense Mutation | AAT,AGT | N,S 326 | XP_011522236.1 | |
XM_011523936.2 | 1374 | Missense Mutation | AAT,AGT | N,S 267 | XP_011522238.1 | |
XM_017024755.1 | 1374 | Missense Mutation | AAT,AGT | N,S 326 | XP_016880244.1 | |
XM_017024756.1 | 1374 | Missense Mutation | AAT,AGT | N,S 326 | XP_016880245.1 | |
XM_017024757.1 | 1374 | Missense Mutation | AAT,AGT | N,S 310 | XP_016880246.1 | |
XM_017024758.1 | 1374 | Missense Mutation | AAT,AGT | N,S 267 | XP_016880247.1 | |
XM_017024759.1 | 1374 | Missense Mutation | AAT,AGT | N,S 267 | XP_016880248.1 |
MYO1C - myosin IC | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |