Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGTGCTTTCCTTTGAGTAATGGT[A/G]TAATATTGTATTTTATGGTGAGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610091 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4522 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4522 - microRNA 4522 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WSB1 - WD repeat and SOCS box containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015626.8 | Intron | NP_056441.6 | ||||
NM_134265.2 | Intron | NP_599027.1 | ||||
XM_005257963.3 | Intron | XP_005258020.1 | ||||
XM_017024433.1 | Intron | XP_016879922.1 | ||||
XM_017024434.1 | Intron | XP_016879923.1 | ||||
XM_017024435.1 | Intron | XP_016879924.1 | ||||
XM_017024436.1 | Intron | XP_016879925.1 |