Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGACCTACTCCCTGACTCGGTT[C/T]GCCATCTACGAGACTGTGCGGGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602375 MIM: 606794 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPL12 - mitochondrial ribosomal protein L12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A10 - solute carrier family 25 member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270888.1 | 574 | Silent Mutation | TTC,TTT | F,F 79 | NP_001257817.1 | |
NM_001270953.1 | 574 | Silent Mutation | TTC,TTT | F,F 79 | NP_001257882.1 | |
NM_012140.4 | 574 | Silent Mutation | TTC,TTT | F,F 79 | NP_036272.2 | |
XM_017024220.1 | 574 | Silent Mutation | TTC,TTT | F,F 36 | XP_016879709.1 |