Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCCACCCATCAGCGCCTTCCCT[A/G]TCAATACGCTCCAGGTAGGAGGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601025 MIM: 611398 MIM: 612128 | ||||||||||||||||||||
Literature Links: |
AP2B1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AP2B1 - adaptor related protein complex 2 beta 1 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GAS2L2 - growth arrest specific 2 like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASL10B - RAS like family 10 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033315.3 | 568 | Missense Mutation | ATC,GTC | I,V 68 | NP_201572.1 | |
XM_017025300.1 | 568 | Missense Mutation | ATC,GTC | I,V 127 | XP_016880789.1 | |
XM_017025301.1 | 568 | Missense Mutation | ATC,GTC | I,V 68 | XP_016880790.1 |