Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCTGGACAACCAAGAAGAAATA[C/G]CGTCATTTTCAGGAGCTGCATCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602384 MIM: 600307 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107984973 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107984973 - atherin-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017025490.1 | 387 | Intron | XP_016880979.1 |
PLD2 - phospholipase D2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243108.1 | 387 | Nonsense Mutation | TAC,TAG | Y,* 104 | NP_001230037.1 | |
NM_002663.4 | 387 | Nonsense Mutation | TAC,TAG | Y,* 104 | NP_002654.3 | |
XM_005256696.2 | 387 | Intron | XP_005256753.1 | |||
XM_017024764.1 | 387 | UTR 5 | XP_016880253.1 |
PSMB6 - proteasome subunit beta 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |