Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTCAACCCAAGCTTTGCGATTA[C/T]AGGTATGGTCTTCGTGGACACTCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601440 | ||||||||||||||||||||
Literature Links: |
C17orf67 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C17orf67 - chromosome 17 open reading frame 67 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DGKE - diacylglycerol kinase epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003647.2 | 642 | Silent Mutation | TAC,TAT | Y,Y 154 | NP_003638.1 | |
XM_011525394.2 | 642 | Silent Mutation | TAC,TAT | Y,Y 172 | XP_011523696.1 | |
XM_011525395.2 | 642 | Silent Mutation | TAC,TAT | Y,Y 172 | XP_011523697.1 | |
XM_011525396.2 | 642 | Silent Mutation | TAC,TAT | Y,Y 172 | XP_011523698.1 | |
XM_017025243.1 | 642 | Silent Mutation | TAC,TAT | Y,Y 154 | XP_016880732.1 | |
XM_017025244.1 | 642 | Silent Mutation | TAC,TAT | Y,Y 172 | XP_016880733.1 | |
XM_017025245.1 | 642 | Silent Mutation | TAC,TAT | Y,Y 172 | XP_016880734.1 |