Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTTCTTAATTCAACTAGGGCAAA[C/T]ATATTTACACAAAGCCACCAGAACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614396 MIM: 607759 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPATCH8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPATCH8 - G-patch domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002909.3 | 5549 | UTR 3 | NP_001002909.1 | |||
NM_001304939.1 | 5549 | UTR 3 | NP_001291868.1 | |||
NM_001304940.1 | 5549 | UTR 3 | NP_001291869.1 | |||
NM_001304941.1 | 5549 | UTR 3 | NP_001291870.1 | |||
NM_001304942.1 | 5549 | UTR 3 | NP_001291871.1 | |||
NM_001304943.1 | 5549 | UTR 3 | NP_001291872.1 | |||
XM_011524557.1 | 5549 | UTR 3 | XP_011522859.1 | |||
XM_011524558.1 | 5549 | UTR 3 | XP_011522860.1 | |||
XM_011524559.2 | 5549 | UTR 3 | XP_011522861.1 | |||
XM_011524561.2 | 5549 | UTR 3 | XP_011522863.1 | |||
XM_017024372.1 | 5549 | UTR 3 | XP_016879861.1 | |||
XM_017024373.1 | 5549 | UTR 3 | XP_016879862.1 | |||
XM_017024374.1 | 5549 | UTR 3 | XP_016879863.1 | |||
XM_017024375.1 | 5549 | UTR 3 | XP_016879864.1 | |||
XM_017024376.1 | 5549 | UTR 3 | XP_016879865.1 | |||
XM_017024377.1 | 5549 | UTR 3 | XP_016879866.1 | |||
XM_017024378.1 | 5549 | UTR 3 | XP_016879867.1 | |||
XM_017024379.1 | 5549 | UTR 3 | XP_016879868.1 | |||
XM_017024380.1 | 5549 | UTR 3 | XP_016879869.1 |
ITGA2B - integrin subunit alpha 2b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |