Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCAGTTCTTTCTACCACACTCC[C/T]GAGGAAGCTCCCATGGACAAAGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616713 | ||||||||||||||||||||
Literature Links: |
PIPOX PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PIPOX - pipecolic acid and sarcosine oxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016518.2 | 459 | Nonsense Mutation | CGA,TGA | R,* 45 | NP_057602.2 | |
XM_017024722.1 | 459 | Intron | XP_016880211.1 |