Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACATGCGGACTGACCTGGCCACAC[C/T]GAGGGAGCTGTCCAAGCAGAGGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601555 MIM: 604631 | ||||||||||||||||||||
Literature Links: |
RND2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RND2 - Rho family GTPase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005440.4 | 336 | Missense Mutation | CCG,CTG | P,L 130 | NP_005431.1 | |
XM_011525316.1 | 336 | Missense Mutation | CCG,CTG | P,L 130 | XP_011523618.1 | |
XM_011525317.2 | 336 | Missense Mutation | CCG,CTG | P,L 102 | XP_011523619.1 |
VAT1 - vesicle amine transport 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |