Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAAAGATCTGGTTTCAGAACAAA[C/T]GCTCCAAGTATAAGAAGCTCCTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601911 | ||||||||||||||||||||
Literature Links: |
DLX4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DLX4 - distal-less homeobox 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001934.3 | 822 | Missense Mutation | CGC,TGC | R,C 97 | NP_001925.2 | |
NM_138281.2 | 822 | Missense Mutation | CGC,TGC | R,C 169 | NP_612138.1 | |
XM_017024291.1 | 822 | Missense Mutation | CGC,TGC | R,C 97 | XP_016879780.1 |