Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAAAAGCTTGGACTCCAAGACGA[G/T]CTCTTCCTTTTATACTGTTCTTATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610730 | ||||||||||||||||||||
Literature Links: |
CCT6B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCT6B - chaperonin containing TCP1 subunit 6B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193529.2 | 1279 | Missense Mutation | GAT,GCT | D,A 396 | NP_001180458.1 | |
NM_001193530.1 | 1279 | Missense Mutation | GAT,GCT | D,A 388 | NP_001180459.1 | |
NM_006584.3 | 1279 | Missense Mutation | GAT,GCT | D,A 433 | NP_006575.2 | |
XM_011524203.2 | 1279 | Missense Mutation | GAT,GCT | D,A 291 | XP_011522505.1 | |
XM_017024023.1 | 1279 | Missense Mutation | GAT,GCT | D,A 367 | XP_016879512.1 | |
XM_017024024.1 | 1279 | Missense Mutation | GAT,GCT | D,A 254 | XP_016879513.1 |