Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTCCTGGGAGTAGAGGTCAGTGA[A/C]GAGAGCTGGCAGCCAGTAGTGGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610904 MIM: 611362 | ||||||||||||||||||||
Literature Links: |
SNF8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNF8 - SNF8, ESCRT-II complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317192.1 | 823 | Missense Mutation | GTC,TTC | V,F 239 | NP_001304121.1 | |
NM_001317193.1 | 823 | Missense Mutation | GTC,TTC | V,F 223 | NP_001304122.1 | |
NM_001317194.1 | 823 | Missense Mutation | GTC,TTC | V,F 136 | NP_001304123.1 | |
NM_007241.3 | 823 | Missense Mutation | GTC,TTC | V,F 240 | NP_009172.2 | |
XM_017024075.1 | 823 | Missense Mutation | GTC,TTC | V,F 136 | XP_016879564.1 |
UBE2Z - ubiquitin conjugating enzyme E2 Z | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |