Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTCGCCAGCTGGTCCTGAGTCT[A/T]CTCAGCCTGGAGCCTGCCCAGCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609799 MIM: 602464 | ||||||||||||||||||||
Literature Links: |
NEK8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NEK8 - NIMA related kinase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178170.2 | 918 | Silent Mutation | CTA,CTT | L,L 238 | NP_835464.1 | |
XM_011524638.2 | 918 | Silent Mutation | CTA,CTT | L,L 262 | XP_011522940.1 | |
XM_011524640.2 | 918 | Silent Mutation | CTA,CTT | L,L 262 | XP_011522942.1 | |
XM_017024499.1 | 918 | Silent Mutation | CTA,CTT | L,L 238 | XP_016879988.1 | |
XM_017024500.1 | 918 | Silent Mutation | CTA,CTT | L,L 199 | XP_016879989.1 | |
XM_017024501.1 | 918 | Silent Mutation | CTA,CTT | L,L 262 | XP_016879990.1 |
TLCD1 - TLC domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRAF4 - TNF receptor associated factor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |